Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gadolinium, plasma standard solution, Specpure™ Gd 1000μg/mL, Thermo Scientific Chemicals

Catalog Number AA13829AD
Click to view available options
Quantity:
50 mL
100 mL
500 mL
12064-62-9, 7732-18-5
Gd2O3
362.50
MFCD00011026
CMIHHWBVHJVIGI-UHFFFAOYSA-N
159427
[O--].[O--].[O--].[Gd+3].[Gd+3]
12064-62-9, 7732-18-5
Gd2O3
362.50
MFCD00011026
CMIHHWBVHJVIGI-UHFFFAOYSA-N
159427
[O--].[O--].[O--].[Gd+3].[Gd+3]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 50 mL
Color Colorless
IUPAC Name gadolinium(3+);oxygen(2-)
Concentration or Composition (by Analyte or Components) Nitric acid: 5.0%; Gadolinium(III) oxide: 0.12%; Water: 94.88%
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Chemical Name or Material Gadolinium
Hazard Category H290-H314-H335
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Hazard Class: 8, Packing Group: III
EINECSNumber 231-714-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit