Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ gag-pol Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144418
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144418 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144418 Supplier Invitrogen™ Supplier No. PA5144418
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

gag-pol Polyclonal Antibody for ELISA, Western Blot

[Gag-Pol polyprotein]: Plays a role in budding and is processed by the viral protease during virion maturation outside the cell. During budding, it recruits, in a PPXY-dependent or independent manner, Nedd4-like ubiquitin ligases that conjugate ubiquitin molecules to Gag-Pol, or to Gag-Pol binding host factors. Interaction with HECT ubiquitin ligases probably links the viral protein to the host ESCRT pathway and facilitates release.
TRUSTED_SUSTAINABILITY

Specifications

Antigen gag-pol
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.76 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene gag-pol
Gene Accession No. P03355
Gene Alias Gag-Pol polyprotein gag-pol
Gene Symbols gag-pol
Host Species Rabbit
Immunogen Recombinant Moloney murine leukemia virus (isolate Shinnick) (MoMLV) POL protein (560-631aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2193424
Target Species Virus
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.