Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GAGE12B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548149
Description
GAGE12B Polyclonal Antibody for Western Blot, IHC (P)

Specifications
GAGE12B | |
Polyclonal | |
Unconjugated | |
GAGE12B | |
AL4; Cancer/testis antigen 4.7; CT4.7; G antigen 12B; G antigen 12B/C/D/E; g antigen 12C/D/E; G antigen 7; GAGE12B; GAGE-12B; GAGE12C; GAGE-12C; GAGE12D; GAGE-12D; GAGE12E; GAGE-12E; GAGE12I; GAGE-12I; GAGE7; GAGE-7; GAGE7B; GAGE-7B; GAGE-8 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
729428 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
A1L429 | |
GAGE12B | |
KLH conjugated synthetic peptide between 21-49 amino acids from the N-terminal region of human GAGE12B. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction