Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GAGE12F/GAGE12G/GAGE12I Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572113
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572113 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572113 Supplier Invitrogen™ Supplier No. PA572113
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GAGE12F/GAGE12G/GAGE12I Polyclonal Antibody for Western Blot

GAGE12F is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAGE12F/GAGE12G/GAGE12I
Applications Western Blot
Classification Polyclonal
Concentration 0.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GAGE12F
Gene Accession No. P0CL80
Gene Alias AL4; Cancer/testis antigen 4.7; CT4.7; G antigen 12F; g antigen 12F/G/I; G antigen 12G; G antigen 12I; G antigen 7; GAGE12F; GAGE-12F; GAGE12G; GAGE-12G; GAGE12I; GAGE-12I; GAGE7; GAGE-7; GAGE7B; GAGE-7B; GAGE-8
Gene Symbols GAGE12F
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human GAGE12F / GAGE12G / GAGE12I.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100008586
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.