Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GAGE12I Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546157
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546157 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546157 Supplier Invitrogen™ Supplier No. PA546157
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: AGQGPKPEAH SQEQGHPQTG CECEDGPDGQ EMDPPNPEEV KTPEEGEKQS Sequence homology: Human: 100%.

This gene belongs to a family of genes that are expressed in a variety of tumors but not in normal tissues, except for the testis. The sequences of the family members are highly related but differ by scattered nucleotide substitutions. The antigenic peptide YYWPRPRRY, which is also encoded by several other family members, is recognized by autologous cytolytic T lymphocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAGE12I
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GAGE12I
Gene Accession No. P0CL82
Gene Alias AL4; cancer/testis antigen 4.7; CT4.7; G antigen 12F; g antigen 12F/G/I; G antigen 12G; G antigen 12I; G antigen 7; G antigen 7B; GAGE12F; GAGE-12F; GAGE12G; GAGE-12G; GAGE12I; GAGE-12I; GAGE7; GAGE-7; GAGE7B; GAGE-7B; GAGE-8
Gene Symbols GAGE12I
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human GAGE12I (aa 66-115).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26748
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.