Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GAGE2B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548522
Description
GAGE2B Polyclonal Antibody for Western Blot
Antigen, recognized on melanoma by autologous cytolytic T-lymphocytes.
Specifications
GAGE2B | |
Polyclonal | |
Unconjugated | |
GAGE2B | |
Cancer/testis antigen 4.2; CT4.2; CT42; g antigen 2A/2B; G antigen 2B; G antigen 2B/2C; G antigen 2C; GAGE2; GAGE-2; GAGE2B; GAGE-2B; GAGE2C; GAGE-2C | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
645037 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q13066 | |
GAGE2B | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human GAGE2B. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction