Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GAGE6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570899
Description
This target displays homology in the following species: Human: 100%.
This gene belongs to a family of genes that are expressed in a variety of tumors but not in normal tissues, except for the testis. The sequences of the family members are highly related but differ by scattered nucleotide substitutions. The antigenic peptide YYWPRPRRY, which is also encoded by several other family members, is recognized by autologous cytolytic T lymphocytes.
Specifications
GAGE6 | |
Polyclonal | |
Unconjugated | |
GAGE6 | |
Cancer/testis antigen 4.6; cancer/testis antigen family 4, member 6; CT4.6; G antigen 6; GAGE6; GAGE-6 | |
Rabbit | |
Affinity chromatography | |
RUO | |
2578 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q13070 | |
GAGE6 | |
synthetic peptide directed towards the C-terminal region of human GAGE6. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction