Learn More
Invitrogen™ GAGE7 Polyclonal Antibody

Description
GAGE7 Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | GAGE7 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | GAGE7 |
| Gene Accession No. | O76087 |
| Gene Alias | AL4; cancer/testis antigen 4.7; cancer/testis antigen family 4, member 7; CT4.7; G antigen 12F; g antigen 12F/G/I; G antigen 12G; G antigen 12I; G antigen 7; GAGE12F; GAGE-12F; GAGE12G; GAGE-12G; GAGE12I; GAGE-12I; GAGE7; GAGE-7; GAGE7B; GAGE-7B; GAGE-8; RP11-959H17.6 |
| Gene Symbols | GAGE7 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.