Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAK Polyclonal Antibody

GAK Polyclonal Antibody
Supplier: Thermo Scientific PA555626
Description
GAK Polyclonal Antibody for IHC

Specifications
GAK | |
Polyclonal | |
Unconjugated | |
GAK | |
auxilin-2; cyclin G associated kinase; cyclin G-associated kinase; cyclin-G-associated kinase; D130045N16Rik; DNAJ26; DNAJC26; GAK | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2580 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O14976 | |
GAK | |
Recombinant protein corresponding to Human GAK. Recombinant protein control fragment (Product #RP-95241). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction