Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAL-4 DNA Binding Domain Mouse anti-Yeast, Clone: 8C-1, Exalpha
SDP

Catalog No. 502102314
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-210-2314 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-210-2314 Supplier Vector Laboratories Supplier No. Z150M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

This antibody detects a 106 kDa protein by Western blot in yeast cell extracts corresponding to amino acids 1-147 of the GAL4 DNA binding region.

GAL-4 DNA Binding Domain Monoclonal antibody specifically detects GAL-4 DNA Binding Domain in Yeast samples. It is validated for Immunoprecipitation, Western Blot

Specifications

Antigen GAL-4 DNA Binding Domain
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 8C-1
Concentration 0.5-2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide; pH
Gene GAL4
Gene Accession No. P04386
Gene Alias antigen NY-CO-27; GAL4; Gal-4; GAL81; galactose-responsive transcription factor GAL4; galectin 4; galectin-4; L-36 lactose-binding protein; L36LBP; Lactose-binding lectin 4; lectin, galactoside binding soluble 4; lectin, galactoside-binding, soluble, 4; LGALS4; Regulatory protein GAL4; YPL248C
Host Species Mouse
Immunogen Hybridoma produced by the fusion of splenocytes from mice immunized with full length GAL4 expressed in E. Coli and mouse myeloma cells.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 855828
Target Species Yeast
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.