Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GAL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562069
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562069 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562069 Supplier Invitrogen™ Supplier No. PA562069
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GPHAVGNHRS FSDKNGLTSK RELRPEDDMK PGSFDRSIPE NNIMRTIIEF LSFLHLKEAG ALDRLLDLPA AASSEDIERS Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 65%.

Endocrine hormone of the central and peripheral nervous systems that binds and activates the G protein-coupled receptors GALR1, GALR2, and GALR3. This small neuropeptide may regulate diverse physiologic functions including contraction of smooth muscle of the gastrointestinal and genitourinary tract, growth hormone and insulin release and adrenal secretion. [Uniprot].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAL
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.40 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Gal
Gene Accession No. P22466
Gene Alias ETL8; GAL; GAL1; Galanin; galanin and GMAP prepropeptide; Galanin message-associated peptide; Galanin peptides; galanin prepropeptide; galanin/GMAP prepropeptide; galanin-message-associated peptide; galanin-related peptide; GAL-GMAP; GALN; GLNN; GMAP; MGC40167; preprogalanin
Gene Symbols Gal
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GAL. Recombinant protein control fragment (Product #RP-101186).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51083
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.