Learn More
Invitrogen™ GAL Polyclonal Antibody

Description
This antibody is predicted to react with bovine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Specifications
Specifications
| Antigen | GAL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | Gal |
| Gene Accession No. | P10683, P22466, P47212 |
| Gene Alias | ETL8; GAL; GAL1; Galanin; galanin and GMAP prepropeptide; Galanin message-associated peptide; Galanin peptides; galanin prepropeptide; galanin/GMAP prepropeptide; galanin-message-associated peptide; galanin-related peptide; GAL-GMAP; GALN; GLNN; GMAP; MGC40167; preprogalanin |
| Gene Symbols | Gal |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.