Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GAL3ST3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543507
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543507 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543507 Supplier Invitrogen™ Supplier No. PA543507
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: AHNTLAYDLG GDNERSPRDD AAYLAGLIRQ VEEVFSLVMI AEYFDESLVL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.

GAL3ST3 is a member of the galactose-3-O-sulfotransferase protein family. It catalyzes sulfonation by transferring a sulfate group to the 3' position of galactose in N-acetyllactosamine in both type 2 (Gal-beta-1-4GlcNAc-R) oligosaccharides and core-2-branched O-glycans, but not on type 1 or core-1-branched structures. This gene, which has also been referred to as GAL3ST2, is different from the GAL3ST2 gene located on chromosome 2 that encodes a related enzyme with distinct tissue distribution and substrate specificities, compared to galactose-3-O-sulfotransferase 3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAL3ST3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GAL3ST3
Gene Accession No. Q96A11
Gene Alias Beta-galactose-3-O-sulfotransferase 3; GAL3ST2; GAL3ST3; Gal3ST-3; galactose 3'-sulfotransferase; galactose-3-O-sulfotransferase 3; gal-beta-1, 3-GalNAc 3'-sulfotransferase 3; galbeta1-3GalNAc 3'-sulfotransferase 3; LOW QUALITY PROTEIN: galactose-3-O-sulfotransferase 3; MGC142112; MGC142114
Gene Symbols GAL3ST3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human GAL3ST3.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89792
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.