Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GAL3ST4 Monoclonal Antibody (4E4)
GREENER_CHOICE

Catalog No. PIMA524391
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524391 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524391 Supplier Invitrogen™ Supplier No. MA524391
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: RLQTAVAELR ARREALAKHC LVGGEASDPK YITDRRFRPF QFGSAKVLGY ILRSGLSPQD QEECERLATP ELQYKDKLDA KQFPPTVSLP LKTSRPLSP.

Catalyzes the transfer of sulfate to beta-1,3-linked galactose residues in O-linked glycoproteins. Good substrates include asialofetuin, Gal-beta-1,3-GalNAc and Gal-beta-1,3 (GlcNAc-beta-1,6)GalNAc.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAL3ST4
Applications ELISA
Classification Monoclonal
Clone 4E4
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene GAL3ST4
Gene Accession No. Q96RP7
Gene Alias 1500031A01Rik; beta-galactose-3-O-sulfotransferase; Beta-galactose-3-O-sulfotransferase 4; beta-galactose-3-O-sulfotransferase, 4; GAL3ST4; Gal3ST-4; Galactose-3-O-sulfotransferase 4; gal-beta-1,3-GalNAc 3'-sulfotransferase; galbeta1-3GalNAc 3'-sulfotransferase; PP6968; RGD1561105
Gene Symbols GAL3ST4
Host Species Mouse
Immunogen GAL3ST4 (NP_078913.3, 388 a.a. ~ 486 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79690
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.