Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GAL4 Monoclonal Antibody (2GV-3)
GREENER_CHOICE

Catalog No. MA3031
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA3031 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA3031 Supplier Invitrogen™ Supplier No. MA3031
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA3-031 recognizes Gal4 DNA Binding Domain corresponding to amino acids 1-147 of the GAL4 DNA binding region.

GAL4 is a transcriptional activator which activates genes necessary for galactose catabolism. The DNA binding domain of GAL4 is contained in the first 74 amino acids and contains a zinc finger sequence motif. However, amino acids 1-147 have been shown to bind to DNA in vitro. While GAL4 (1-74) bind DNA specifically, it is with a lower affinity than the GAL4 (1-147). GAL4 (1-147) binds to a specific 17 base pair nucleotide sequence, but alone is insufficient to activate transcription. Either of the two acidic activating regions, activating region I (amino acids 148-238) or activating region II (amino acids 768-881), must be joined to activate transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAL4
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2GV-3
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.05% sodium azide
Gene GAL4
Gene Accession No. P04386
Gene Alias antigen NY-CO-27; GAL4; Gal-4; GAL81; galactose-responsive transcription factor GAL4; galectin 4; galectin-4; L-36 lactose-binding protein; L36LBP; Lactose-binding lectin 4; lectin, galactoside binding soluble 4; lectin, galactoside-binding, soluble, 4; LGALS4; Regulatory protein GAL4; YPL248C
Gene Symbols GAL4
Host Species Mouse
Immunogen Gal4 DNA Binding Domain aa1-147.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 855828
Target Species Yeast
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.