Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Galactocerebroside, Mouse, Alexa Fluor 647, Clone: mGalC,

Catalog No. MAB342AF647 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Alexa Fluor 647
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MAB342AF647 100 μL Alexa Fluor 647
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB342AF647 Supplier MilliporeSigma™ Supplier No. MAB342AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Galactocerebroside (GalCer or GalC) is the most frequently found glycosphingolipid in animals. GalCer consist of a ceramide linked to a single galactose residue at the 1-hydroxyl moiety. Cerebrosides are concentrated in nervous tissues where they increase from about 0.02% of the dry weight in human fetal brain to 2% in gray matter and 12% in white matter of adult brain. GalCer is a specific cell-surface antigenic marker for oligodendrocytes and schwann cells. GalCer is the largest single component of the myelin sheath of nerves. GalCer synthesis therefore serves as a measurement of myelin formation or remyelination. Krabbe disease is an autosomal recessive sphingolipidosis caused by deficient activity of the lysosomal hydrolase galactosylceramide beta-galactosidase (GALC). GALC mediates the degradation of GalCer and other terminal beta-Gal-containing sphingolipids, including psychosine (galactosylsphingosine). Increased psychosine levels are believed to lead to widespread destruction of oligodendroglia in the CNS and to subsequent demyelination.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Galactocerebroside
Applications Immunocytochemistry
Classification Monoclonal
Clone mGalC
Concentration Please refer to lot specific datasheet.
Conjugate Alexa Fluor 647
Formulation Purified mouse IgG3 Alexa Fluor™ 647 conjugate in PBS with 1.5% BSA and 0.09% Sodium Azide.
Host Species Mouse
Immunogen Synaptic plasma membranes from bovine hippocampus (Ranscht, B. et al. (1982) Proc. Natl. Acad. Sci. U. S. A. 79(8):2709-2713).
Purification Method Protein A Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Galactocerebroside (GalC), sulfatide, psychosine and other galactolipids. Cross-reacts with the sulfatide ester of GalC, but to a 16-fold lesser extent. No cross-reactivity with sphingosine, ceramide, mixed ganglioside or glucocerebroside. Binds specifically to the surface of oligodendrocytes and Schwann cells (Ranscht, B. et al. (1982) Proc. Natl. Acad. Sci. U. S. A. 79(8):2709-2713).
Target Species Bovine
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Isotype IgG3
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.