Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galanin Mouse anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501979992
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-197-9992 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-197-9992 Supplier Bioss Supplier No. BS0017M
Add to Cart
Add to Cart

Mouse Polyclonal Antibody

The activity of this receptor for the hormone galanin is mediated by G proteins of the Gi/Go family that inhibit adenylate cyclase activity. GALR1 is widely expressed in the brain and spinal cord, as well as in peripheral sites such as the colon, small intestine and heart. ESTs have been isolated from nerve and fetal lung/testis/B-cell libraries.

Specifications

Antigen Galanin
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Gal
Gene Accession No. P22466
Gene Alias ETL8; GAL; GAL1; Galanin; galanin and GMAP prepropeptide; Galanin message-associated peptide; Galanin peptides; galanin prepropeptide; galanin/GMAP prepropeptide; galanin-message-associated peptide; galanin-related peptide; GAL-GMAP; Galn; GLNN; GMAP; MGC40167
Gene Symbols Gal
Host Species Mouse
Immunogen KLH conjugated synthetic peptide derived from human Galanin.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51083
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.