Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galanin R1/GALR1 Antibody (613207) [DyLight 680], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10353Y
Description
Galanin R1/GALR1 Monoclonal antibody specifically detects Galanin R1/GALR1 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
Galanin R1/GALR1 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
613207 | |
Flow Cytometry, CyTOF-ready | |
galanin receptor 1, galanin receptor type 1, GALNRGALNR1GAL1-R, GALR-1 | |
NS0 mouse myeloma cell line transfected with human Galanin R1/GALR1, Accession # P47211 | |
0.1 mL | |
GPCR | |
2587 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction