Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galanin R2/GALR2 Antibody (616520) [DyLight 488], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6544K
Description
Galanin R2/GALR2 Monoclonal antibody specifically detects Galanin R2/GALR2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
Galanin R2/GALR2 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, CyTOF | |
616520 | |
Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
GAL2-R, galanin receptor 2, galanin receptor type 2, GALNR2MGC125984, GALR-2, MGC125983 | |
NS0 mouse myeloma cell line transfected with human Galanin R2, Accession # O43603 | |
0.1 mL | |
GPCR | |
8811 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction