Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GALC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599988
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599988 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599988 Supplier Invitrogen™ Supplier No. PA599988
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Galc.

This gene encodes a lysosomal protein which hydrolyzes the galactose ester bonds of galactosylceramide, galactosylsphingosine, lactosylceramide, and monogalactosyldiglyceride. Mutations in this gene have been associated with Krabbe disease, also known as globoid cell leukodystrophy. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GALC
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Galc
Gene Accession No. P54803, P54818
Gene Alias 2310068B06Rik; 3.2.1.46; A930008M05Rik; AW212969; AW413532; beta-galactosidase; casein kinase 2, beta polypeptide pseudogene; Gacy; Galactocerebrosidase; galactocerebroside beta-galactosidase; Galactosylceramidase; galactosylceramidase (Krabbe disease); galactosylceramide beta-galactosidase; galactosylceraminidase; GALC; GALCERase; testis tissue sperm-binding protein Li 88E; testis tissue sperm-binding protein Li 89A; twi; twitcher
Gene Symbols Galc
Host Species Rabbit
Immunogen A synthesized peptide derived from human GALC(Accession P54803), corresponding to amino acid residues W342-S392.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14420, 2581, 314360
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.