Learn More
Invitrogen™ GALC Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109753
Description
Immunogen sequence: GGAYVLDDSD GLGREFDGIG AVSGGGATSR LLVNYPEPYR SQILDYLFKP NFGASLHILK VEIGGDGQTT DGTEPSHMHY ALDENYFRGY EWWLMKEAKK RNPNITLIGL PWSFPGWLGK GFDWPYVNLQ LTAYYVVTWI VGAKRYHDLD IDYIGIWNER SYNANYIKIL RKMLNYQGLQ RVKIIASDNL WESISASMLL DAELFKVVDV IGAHYPGTHS AKDAKLTGKK LWSSEDFSTL NSDMGAGCWG RILNQNYING Y.
This gene encodes a lysosomal protein which hydrolyzes the galactose ester bonds of galactosylceramide, galactosylsphingosine, lactosylceramide, and monogalactosyldiglyceride. Mutations in this gene have been associated with Krabbe disease, also known as globoid cell leukodystrophy. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
GALC | |
Polyclonal | |
Unconjugated | |
Galc | |
2310068B06Rik; 3.2.1.46; A930008M05Rik; AW212969; AW413532; beta-galactosidase; casein kinase 2, beta polypeptide pseudogene; Gacy; Galactocerebrosidase; galactocerebroside beta-galactosidase; Galactosylceramidase; galactosylceramidase (Krabbe disease); galactosylceramide beta-galactosidase; galactosylceraminidase; GALC; GALCERase; testis tissue sperm-binding protein Li 88E; testis tissue sperm-binding protein Li 89A; twi; twitcher | |
Rabbit | |
Affinity Chromatography | |
RUO | |
14420, 2581, 314360 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1.37 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P54803, P54818 | |
Galc | |
Recombinant fusion protein containing a sequence corresponding to amino acids 40-300 of human GALC (NP_000144.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.