Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GALE Antibody (6G10), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259421H
This item is not returnable.
View return policy
Description
GALE Monoclonal antibody specifically detects GALE in Human samples. It is validated for Western Blot, ELISASpecifications
GALE | |
Monoclonal | |
HRP | |
EC 5.1.3, EC 5.1.3.2, FLJ95174, FLJ97302, Galactowaldenase, short chain dehydrogenase/reductase family 1E, member 1, UDP-, UDP galactose-4'-epimerase, UDP-galactose 4-epimerase, UDP-galactose-4-epimerase, UDP-glucose 4-epimerase | |
Recombinant human GALE (1-348aa) purified from E. coli | |
0.1 mL | |
Lipid and Metabolism | |
2582 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA | |
6G10 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction