Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GALE Antibody (6G10), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259421PCP
Description
GALE Monoclonal antibody specifically detects GALE in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| GALE | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry | |
| 6G10 | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| EC 5.1.3, EC 5.1.3.2, FLJ95174, FLJ97302, Galactowaldenase, short chain dehydrogenase/reductase family 1E, member 1, UDP-, UDP galactose-4'-epimerase, UDP-galactose 4-epimerase, UDP-galactose-4-epimerase, UDP-glucose 4-epimerase | |
| Recombinant human GALE (1-348aa) purified from E. coli | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 2582 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction