Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GALE Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144982
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144982 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144982 Supplier Invitrogen™ Supplier No. PA5144982
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RSVEFEEMDI LDQGALQRLF KKYSFMAVIH FAGLKAVGES VQKPLDYYRV NLTGTIQLLE IMKAHGVKNL VFSSSATVYG NPQYLPLDEA HPTGGCTNPY GKSKFFIEEM IRDLCQADKT WNAVL Antigen sequence identity: Mouse 94%; Rat 91%.

This gene encodes UDP-galactose-4-epimerase which catalyzes two distinct but analogous reactions: the epimerization of UDP-glucose to UDP-galactose, and the epimerization of UDP-N-acetylglucosamine to UDP-N-acetylgalactosamine. The bifunctional nature of the enzyme has the important metabolic consequence that mutant cells (or individuals) are dependent not only on exogenous galactose, but also on exogenous N-acetylgalactosamine as a necessary precursor for the synthesis of glycoproteins and glycolipids. Mutations in this gene result in epimerase-deficiency galactosemia, also referred to as galactosemia type 3, a disease characterized by liver damage, early-onset cataracts, deafness and mental retardation, with symptoms ranging from mild ('peripheral' form) to severe ('generalized' form). Multiple alternatively spliced transcripts encoding the same protein have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GALE
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GALE
Gene Accession No. Q14376
Gene Alias 2310002A12Rik; AI323962; EGK_00373; galactose-4-epimerase, UDP; galactose-4-epimerase, UDP-; galactowaldenase; Gale; SDR1E1; short chain dehydrogenase/reductase family 1E, member 1; UDP galactose-4'-epimerase; UDP-galactosamine 4-epimerase; UDP-galactose 4-epimerase; UDP-galactose-4-epimerase; UDP-GalNAc 4-epimerase; UDP-GlcNAc 4-epimerase; UDP-glucose 4-epimerase; UDP-N-acetylgalactosamine 4-epimerase; UDP-N-acetylglucosamine 4-epimerase
Gene Symbols GALE
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 58-182. Recombinant protein control fragment (Product #RP-109862).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2582
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.