Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galectin-1 Antibody (001), DyLight 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289387UV
Description
Galectin-1 Monoclonal specifically detects Galectin-1 in Human samples. It is validated for Western Blot.Specifications
Galectin-1 | |
Monoclonal | |
DyLight 350 | |
14 kDa laminin-binding protein, 14 kDa lectin, Beta-galactoside-binding lectin L-14-I, DKFZp686E23103, gal-1, GAL1, Galaptin, galectin 1, galectin-1, GBP, HBL, HLBP14, HPL, Lactose-binding lectin 1, Lectin galactoside-binding soluble 1, lectin, galactoside-binding, soluble, 1, Putative MAPK-activating protein PM12, S-Lac lectin 1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Galectin-1 (NP_002296.1; Met1-Asp135). | |
0.1 mL | |
Apoptosis, Biologically Active Proteins | |
3956 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
1 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction