Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galectin-1 Antibody (RM0081-9J26), Alexa Fluor™ 488, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP121721AF488
Description
Galectin-1 Monoclonal specifically detects Galectin-1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
Galectin-1 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
LGALS1 | |
Mouse recombinant Galectin-1 | |
0.1 mL | |
Primary | |
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant Galectin-1. | |
Store at 4C in the dark. | |
IgG2 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
RM0081-9J26 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen | |
14 kDa laminin-binding protein, 14 kDa lectin, Beta-galactoside-binding lectin L-14-I, DKFZp686E23103, gal-1, GAL1, Galaptin, galectin 1, galectin-1, GBP, HBL, HLBP14, HPL, Lactose-binding lectin 1, Lectin galactoside-binding soluble 1, lectin, galactoside-binding, soluble, 1, Putative MAPK-activating protein PM12, S-Lac lectin 1 | |
Rat | |
Protein A or G purified | |
Apoptosis, Biologically Active Proteins | |
3956 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction