Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galectin-1 Antibody (RM0081-9J26) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50 - $560.50
Specifications
| Antigen | Galectin-1 |
|---|---|
| Clone | RM0081-9J26 |
| Concentration | LYOPH |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
Description
Galectin-1 Monoclonal specifically detects Galectin-1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
| Galectin-1 | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| 14 kDa laminin-binding protein, 14 kDa lectin, Beta-galactoside-binding lectin L-14-I, DKFZp686E23103, gal-1, GAL1, Galaptin, galectin 1, galectin-1, GBP, HBL, HLBP14, HPL, Lactose-binding lectin 1, Lectin galactoside-binding soluble 1, lectin, galactoside-binding, soluble, 1, Putative MAPK-activating protein PM12, S-Lac lectin 1 | |
| LGALS1 | |
| Primary | |
| This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant Galectin-1. |
| RM0081-9J26 | |
| Immunohistochemistry (Frozen), Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| 3956 | |
| Mouse recombinant Galectin-1 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title