Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galectin-1 Antibody (RM0081-9J26), FITC, Novus Biologicals™
SDP

Rat Monoclonal Antibody

Supplier:  Novus Biologicals NBP121721F

Catalog No. NBP121721F


Only null left
Add to Cart

Description

Description

Galectin-1 Monoclonal specifically detects Galectin-1 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications

Specifications

Galectin-1
Monoclonal
FITC
PBS with 0.05% Sodium Azide
LGALS1
Mouse recombinant Galectin-1
0.1 mL
Primary
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant Galectin-1.
Store at 4C in the dark.
IgG2
Immunohistochemistry, Immunohistochemistry (Frozen)
RM0081-9J26
Immunohistochemistry, Immunohistochemistry-Frozen
14 kDa laminin-binding protein, 14 kDa lectin, Beta-galactoside-binding lectin L-14-I, DKFZp686E23103, gal-1, GAL1, Galaptin, galectin 1, galectin-1, GBP, HBL, HLBP14, HPL, Lactose-binding lectin 1, Lectin galactoside-binding soluble 1, lectin, galactoside-binding, soluble, 1, Putative MAPK-activating protein PM12, S-Lac lectin 1
Rat
Protein A or G purified
Apoptosis, Biologically Active Proteins
3956
Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.