Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Galectin 1 Polyclonal Antibody
Rabbit Polyclonal Antibody
$178.40 - $482.00
Specifications
Antigen | Galectin 1 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119132
|
Invitrogen™
PA5119132 |
200 μL |
Each for $482.00
|
|
|||||
PIPA5121970
|
Invitrogen™
PA5121970 |
20 μL |
Each for $178.40
|
|
|||||
Description
Galectin 1 Polyclonal Antibody for Western Blot, IHC (P)
Galectin-1 belongs to a large family of carbohydrate-binding proteins called lectins. Galectin-1can be either monomeric or homodimeric and is found in a wide variety of cells and tissue types. Galectin-1 can control cell growth, proliferation, induce apoptosis of activated T cells while it can also modulate cytokine secretion or inhibit pro-inflammatory cytokine production. Galectin-1 plays an important role in acute and chronic inflammation.Specifications
Galectin 1 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human, Mouse | |
LGALS1 | |
14 kDa laminin-binding protein; 14 kDa lectin; AA410090; beta-galactoside binding protein; beta-galactoside-binding lectin; beta-galactoside-binding lectin L-14-I; beta-galactoside-binding protein 14kDa; DKFZp686E23103; GAL1; GAL-1; Galaptin; Galbp; galectin 1; galectin 1; LGALS1; Galectin1; galectin-1; galectin-1-like protein; GBP; HBL; H-Galectin-1; HLBP14; HPL; L14; L-14.5; Lactose-binding lectin 1; Lect14; Lectin galactoside-binding soluble 1; lectin, galactose binding, soluble 1; lectin, galactoside binding soluble 1; lectin, galactoside-binding, soluble, 1; lectin, galactoside-binding, soluble, 1 (galectin 1); Lgals1; LGAS1; OPG-1; pm12; Putative MAPK-activating protein PM12; RL 14.5; similar to galactose-binding lectin; S-Lac lectin 1; unnamed protein product | |
LGALS1 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
P09382, P16045 | |
16852, 3956 | |
Recombinant protein Galectin 1. The antigen corresponds to amino acid range 1-135 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title