Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galectin-1 Rabbit anti-Human, Clone: 8, Novus Biologicals™

Rabbit Monoclonal Antibody
$342.00 - $524.00
Specifications
Antigen | Galectin-1 |
---|---|
Clone | 8 |
Dilution | ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
Galectin-1 Monoclonal specifically detects Galectin-1 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
Galectin-1 | |
ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
14 kDa laminin-binding protein, 14 kDa lectin, Beta-galactoside-binding lectin L-14-I, DKFZp686E23103, gal-1, GAL1, Galaptin, galectin 1, galectin-1, GBP, HBL, HLBP14, HPL, Lactose-binding lectin 1, Lectin galactoside-binding soluble 1, lectin, galactoside-binding, soluble, 1, Putative MAPK-activating protein PM12, S-Lac lectin 1 | |
Recombinant Human Galectin-1 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
8 | |
ELISA | |
Unconjugated | |
Rabbit | |
Apoptosis, Biologically Active Proteins | |
0.2 um filtered solution in PBS | |
3956 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title