Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Galectin 10 Monoclonal Antibody (561603)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524251
Description
In direct ELISAs, no cross-reactivity with recombinant human Galectin-1, -2, -3, -4, -7, -8, or -9/Ecalectin is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Regulates immune responses through the recognition of cell-surface glycans. Essential for the anergy and suppressive function of CD25-positive regulatory T-cells (Treg).
Specifications
Galectin 10 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q05315 | |
CLC | |
E. coli-derived recombinant human Galectin-10 Met1-Arg142. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Western Blot, Immunocytochemistry | |
561603 | |
Unconjugated | |
CLC | |
Charcot-Leyden crystal galectin; Charcot-Leyden crystal protein; CLC; EC 3.1.1.5; Eosinophil lysophospholipase; GAL10; Gal-10; galectin 10; galectin-10; lectin, galactoside-binding, soluble, 10; LGALS10; LGALS10A; LPPL_HUMAN; lysolecithin acylhydrolase; MGC149659 | |
Mouse | |
Protein A/G | |
RUO | |
1178 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction