Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Galectin 10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100747
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100747 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100747 Supplier Invitrogen™ Supplier No. PA5100747
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LPPL.

Regulates immune responses through the recognition of cell-surface glycans. Essential for the anergy and suppressive function of CD25-positive regulatory T-cells (Treg).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Galectin 10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CLC
Gene Accession No. P97400, Q05315
Gene Alias Charcot-Leyden crystal galectin; Charcot-Leyden crystal protein; CLC; EC 3.1.1.5; Eosinophil lysophospholipase; GAL10; Gal-10; galectin 10; galectin-10; lectin, galactoside-binding, soluble, 10; LGALS10; LGALS10A; LPPL_HUMAN; lysolecithin acylhydrolase; MGC149659
Gene Symbols CLC
Host Species Rabbit
Immunogen A synthesized peptide derived from human CLC(Accession Q05315), corresponding to amino acid residues T19-Y69.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1178, 12735
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.