Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galectin 13 Antibody (PP13/1165), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$243.50 - $487.50

Specifications

Antigen Galectin 13
Clone PP13/1165
Concentration 0.2 mg/mL
Dilution Flow Cytometry 0.5 - 1 ug/million cells, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500, SDS-Page
Applications Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB25316820
SDP
View Documents
Novus Biologicals
NBP25316820UG
20 μg
Each for $243.50
Only null left
Add to Cart
 
NB253168100
SDP
View Documents
Novus Biologicals
NBP253168100UG
100 μg
Each for $487.50
Only null left
Add to Cart
 
NB253168200
SDP
View Documents
Novus Biologicals
NBP253168200UG
200 μg N/A N/A N/A
Description

Description

Galectin 13 Monoclonal specifically detects Galectin 13 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Galectin 13
0.2 mg/mL
Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page
Unconjugated
Mouse
Human
beta-galactoside-binding lectin, EC 3.1.1.5, gal-13, GAL13, galectin-13, lectin, galactoside-binding, soluble, 13, PLAC8galectin 13, Placental protein 13, Placental tissue protein 13, PP13galactoside-binding soluble lectin 13
LGALS13
IgG2b
Protein A or G purified
PP13/1165
Flow Cytometry 0.5 - 1 ug/million cells, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500, SDS-Page
Monoclonal
Purified
RUO
1mM PBS with 0.05% BSA with 0.05% Sodium Azide
29124
Recombinant human Galectin-13 protein fragment (aa23-134) (exact sequence is proprietary)
Primary
Store at 4C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.