Learn More
Invitrogen™ Galectin 14 Monoclonal Antibody (511124)

Description
In direct ELISAs, no cross-reactivity with recombinant human Galectin-1, -2, -3, -4, -7, -8, -10, or recombinant mouse Galectin-9 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | Galectin 14 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 511124 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | LGALS14 |
| Gene Accession No. | Q8TCE9 |
| Gene Alias | Charcot-Leyden crystal protein 2; CLC2; gal-14; galectin 14; galectin-14; lectin, galactoside binding soluble 14; lectin, galactoside-binding, soluble, 14; LGALS14; MGC22235; Placental protein 13-like; placental protein 13-like protein; PPL13 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.