Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Galectin 14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5139873
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: QFRLHFGHPA IMNSRVFGIW RYEEKCYYLP FEDGKPF.
Galectin-14 is a member of the galectin family of proteins, which are characterized by their affinity for beta-galactoside sugars and involvement in various biological processes including cell-cell adhesion, immune response modulation, and apoptosis. The gene encoding Galectin-14, LGALS14, is located on chromosome 19q13.2. Galectin-14 is predominantly expressed in eosinophils and has been identified to play a role in allergic response and inflammation. It is known to participate in the modulation of immune responses, particularly asthma and other allergic conditions, by interacting with glycoproteins on cell surfaces. The expression of Galectin-14 is elevated during allergic reactions and this molecule is considered a potential biomarker for eosinophilic activity in such conditions. There is ongoing research into the exact mechanisms of Galectin-14 in immune response modulation, which might provide insights into novel therapeutic approaches for managing allergic diseases.
Specifications
| Galectin 14 | |
| Polyclonal | |
| Unconjugated | |
| LGALS14 | |
| Charcot-Leyden crystal protein 2; CLC2; gal-14; galectin 14; galectin-14; lectin, galactoside binding soluble 14; lectin, galactoside-binding, soluble, 14; LGALS14; MGC22235; Placental protein 13-like; placental protein 13-like protein; PPL13 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56891 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TCE9 | |
| LGALS14 | |
| Recombinant protein corresponding to Human LGALS14. Recombinant protein control fragment (Product #RP-109081). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction