Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Galectin 14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598741
Description
Galectin 14 Polyclonal Antibody for Western Blot, ELISA
Galectin-14 is a member of the galectin family of proteins, which are characterized by their affinity for beta-galactoside sugars and involvement in various biological processes including cell-cell adhesion, immune response modulation, and apoptosis. The gene encoding Galectin-14, LGALS14, is located on chromosome 19q13.2. Galectin-14 is predominantly expressed in eosinophils and has been identified to play a role in allergic response and inflammation. It is known to participate in the modulation of immune responses, particularly asthma and other allergic conditions, by interacting with glycoproteins on cell surfaces. The expression of Galectin-14 is elevated during allergic reactions and this molecule is considered a potential biomarker for eosinophilic activity in such conditions. There is ongoing research into the exact mechanisms of Galectin-14 in immune response modulation, which might provide insights into novel therapeutic approaches for managing allergic diseases.
Specifications
Galectin 14 | |
Polyclonal | |
Unconjugated | |
LGALS14 | |
Charcot-Leyden crystal protein 2; CLC2; gal-14; galectin 14; galectin-14; lectin, galactoside binding soluble 14; lectin, galactoside-binding, soluble, 14; LGALS14; MGC22235; Placental protein 13-like; placental protein 13-like protein; PPL13 | |
Rabbit | |
Protein G | |
RUO | |
56891 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
4.29 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q8TCE9 | |
LGALS14 | |
Recombinant Human Placental protein 13-like protein (31-139AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction