Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galectin-2 Antibody (2509A) [DyLight 350], Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB1153D
Description
Galectin-2 Monoclonal antibody specifically detects Galectin-2 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOFSpecifications
Galectin-2 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
2509A | |
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
Beta-galactoside-binding lectin L-14-II, Gal-2, galectin 2, galectin-2, HL14gal-2, Lactose-binding lectin 2, lectin, galactoside-binding, soluble, 2, MGC75071, S-Lac lectin 2 | |
E. coli-derived human Galectin-2, Met1-Glu132, Accession # P05162 | |
0.1 mL | |
Apoptosis, Cancer, Cellular Markers, Tumor Suppressors | |
3957 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction