Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Galectin-2 Antibody (2509A), mFluor Violet 610 SE, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB1153MFV610
Description
Galectin-2 Monoclonal antibody specifically detects Galectin-2 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
| Galectin-2 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Apoptosis, Cancer, Cellular Markers, Tumor Suppressors | |
| 3957 | |
| Store at 4C in the dark. | 
| Western Blot, Flow Cytometry, CyTOF | |
| 2509A | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| Beta-galactoside-binding lectin L-14-II, Gal-2, galectin 2, galectin-2, HL14gal-2, Lactose-binding lectin 2, lectin, galactoside-binding, soluble, 2, MGC75071, S-Lac lectin 2 | |
| E. coli-derived human Galectin-2, Met1-Glu132, Accession # P05162 | |
| RUO | |
| Primary | |
| Human | |
| IgG | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction