Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galectin-2 Antibody (OTI6A8), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271123AF532
Description
Galectin-2 Monoclonal specifically detects Galectin-2 in Human samples. It is validated for Western Blot.Specifications
Galectin-2 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
LGALS2 | |
Full length recombinant protein of human Galectin-2 (NP_006489) produced in E.coli. | |
Primary | |
Human | |
IgG1 |
Western Blot | |
OTI6A8 | |
Western Blot | |
Beta-galactoside-binding lectin L-14-II, Gal-2, galectin 2, galectin-2, HL14gal-2, Lactose-binding lectin 2, lectin, galactoside-binding, soluble, 2, MGC75071, S-Lac lectin 2 | |
Mouse | |
0.1 mL | |
3957 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction