Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Galectin 2 Monoclonal Antibody (C6)
Mouse Monoclonal Antibody
$213.50 - $522.50
Specifications
Antigen | Galectin 2 |
---|---|
Clone | C6 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541591
|
Invitrogen™
MA541591 |
200 μL |
Each for $522.50
|
|
|||||
PIMA543814
|
Invitrogen™
MA543814 |
20 μL |
Each for $213.50
|
|
|||||
Description
Galectin 2 Monoclonal Antibody for Western Blot, IHC (P)
The protein encoded by this gene is a soluble beta-galactoside binding lectin. The encoded protein is found as a homodimer and can bind to lymphotoxin-alpha. A single nucleotide polymorphism in an intron of this gene can alter the transcriptional level of the protein, with a resultant increased risk of myocardial infarction.Specifications
Galectin 2 | |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P05162 | |
3957 | |
Recombinant protein Galectin 2. The antigen corresponds to amino acid range 1-132 of the target protein. | |
Antibody |
C6 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG2b κ | |
Human | |
LGALS2 | |
2200008F12Rik; AI324147; Beta-galactoside-binding lectin L-14-II; gal-2; galectin 2; galectin-2; HL14; lactose-binding lectin 2; lectin galactoside-binding soluble 2 (galectin 2); lectin, galactose-binding, soluble 2; lectin, galactoside binding soluble 2; lectin, galactoside-binding, soluble, 2; lectin, galactoside-binding, soluble, 2 (galectin 2); LEG2; Lgals2; S-Lac lectin 2 | |
LGALS2 | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title