Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Galectin 2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551955
Description
Immunogen sequence: LEVKNMDMKP GSTLKITGSI ADGTDGFVIN LGQGTDKLNL HFNPRFSEST IVCNSLDGSN WGQEQREDHL CFSPGSEVKF TVTFESDKFK VKLPDGHELT FPNRLGHSHL SYLSVRGGFN MSSFKL Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 68%.
The protein encoded by this gene is a soluble beta-galactoside binding lectin. The encoded protein is found as a homodimer and can bind to lymphotoxin-alpha. A single nucleotide polymorphism in an intron of this gene can alter the transcriptional level of the protein, with a resultant increased risk of myocardial infarction.
Specifications
Galectin 2 | |
Polyclonal | |
Unconjugated | |
LGALS2 | |
2200008F12Rik; AI324147; Beta-galactoside-binding lectin L-14-II; gal-2; galectin 2; galectin-2; HL14; lactose-binding lectin 2; lectin galactoside-binding soluble 2 (galectin 2); lectin, galactose-binding, soluble 2; lectin, galactoside binding soluble 2; lectin, galactoside-binding, soluble, 2; lectin, galactoside-binding, soluble, 2 (galectin 2); LEG2; Lgals2; S-Lac lectin 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3957 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P05162 | |
LGALS2 | |
Recombinant protein corresponding to Human Galectin 2. Recombinant protein control fragment (Product #RP-91491). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction