Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galectin-3 Antibody (078), Alexa Fluor™ 350

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289384AF350
Description
Galectin-3 Monoclonal specifically detects Galectin-3 in Human samples. It is validated for Western Blot.Specifications
Galectin-3 | |
Monoclonal | |
Alexa Fluor 350 | |
Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, Gal-3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, GALIG, IgE-binding protein, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, MAC-2, Mac-2 antigen, MAC2GAL3 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Galectin-3 (P17931; Met1-Ile250). | |
0.1 mL | |
Apoptosis, Biologically Active Proteins, Cancer, Cellular Markers, Neuroscience, Tumor Suppressors | |
3958 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
78 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction