Learn More
Galectin-3 Antibody (194801), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB11542MFV500
Description
Galectin-3 Monoclonal antibody specifically detects Galectin-3 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
| Galectin-3 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Apoptosis, Biologically Active Proteins, Cancer, Cellular Markers, Neuroscience, Tumor Suppressors | |
| 3958 | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry, CyTOF | |
| 194801 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, Gal-3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, GALIG, IgE-binding protein, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, MAC-2, Mac-2 antigen, MAC2GAL3 | |
| E. coli-derived recombinant human Galectin-3, Ala2-Ile250, Accession # P17931 | |
| RUO | |
| Primary | |
| Human | |
| IgG2b |