Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galectin-3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP216589
Description
Galectin-3 Polyclonal specifically detects Galectin-3 in Human, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Sandwich ELISA, Knockout Validated.Specifications
Galectin-3 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7), 20% Glycerol with 0.025% Proclin 300 | |
Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, Gal-3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, GALIG, IgE-binding protein, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, MAC-2, Mac-2 antigen, MAC2GAL3 | |
Rabbit | |
26 kDa | |
0.1 mL | |
Apoptosis, Cancer, Cellular Markers, Neuroscience, Tumor Suppressors | |
3958 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ELISA, Immunoassay | |
1mg/mL | |
Western Blot 1:5000-1:20000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Sandwich ELISA, Knockout Validated | |
P17931 | |
LGALS3 | |
Recombinant protein encompassing a sequence within the center region of human Galectin 3. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human, Rat, Canine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction