Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galectin-3 Antibody (OTI1C7), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271124F
Description
Galectin-3 Monoclonal specifically detects Galectin-3 in Human, Mouse samples. It is validated for Immunohistochemistry.Specifications
Galectin-3 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
LGALS3 | |
Full length recombinant protein of human Galectin-3 (NP_002297) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG |
Immunohistochemistry | |
OTI1C7 | |
Immunohistochemistry | |
Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, Gal-3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, GALIG, IgE-binding protein, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, MAC-2, Mac-2 antigen, MAC2GAL3 | |
Mouse | |
Affinity Purified | |
Apoptosis, Biologically Active Proteins, Cancer, Cellular Markers, Neuroscience, Tumor Suppressors | |
3958.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction