Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech Galectin 3 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Catalog No. 500P246BT50
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
500P246BT50 50 μg
500P246BT25 25 μg
50-271-8574 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 500P246BT50 Supplier PeproTech Supplier No. 500P246BT50UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: ADNFSLHDAL SGSGNPNPQG WPGAWGNQPA GAGGYPGASY PGAYPGQAPP GAYPGQAPPG AYHGAPGAYP GAPAPGVYPG PPSGPGAYPS SGQPSAPGAY PATGPYGAPA GPLIVPYNLP LPGGVVPRML ITILGTVKPN ANRIALDFQR GNDVAFHFNP RFNENNRRVI VCNTKLDNNW GREERQSVFP FESGKPFKIQ VLVEPDHFKV AVNDAHLLQY NHRVKKLNEI SKLGISGDID LTSASYTMI. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Galectin-3. Anti-Human Galectin-3-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hGalectin-3 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human Galectin-3 (500-P246) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hGalectin-3. Western Blot: To detect hGalectin-3 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hGalectin-3 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P246BT-1MG will be provided as 2 x 500 μg

Galactose-specific lectin which binds IgE. May mediate with the alpha-3, beta-1 integrin the stimulation by CSPG4 of endothelial cells migration. Together with DMBT1, required for terminal differentiation of columnar epithelial cells during early embryogenesis. In the nucleus: acts as a pre-mRNA splicing factor. Involved in acute inflammatory responses including neutrophil activation and adhesion, chemoattraction of monocytes macrophages, opsonization of apoptotic neutrophils, and activation of mast cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Galectin 3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Biotin
Formulation PBS with no preservative
Gene LGALS3
Gene Accession No. P17931
Gene Alias 35 kDa lectin; advanced glycation end-product receptor 3; AGE-R3; beta-galactosides-binding lectin; carbohydrate-binding protein 35; CBP 35; CBP30; CBP-30; CBP35; epsilon BP; gal3; gal-3; galactose-specific lectin 3; Galactoside-binding protein; GALBP; galectin 3; Galectin3; galectin-3; GALIG; GBP; IgE binding protein; igE-binding protein; L-29; L31; L-31; L-34; L-34 galactoside-binding lectin; Laminin-binding protein; lectin galactoside-binding soluble 3; lectin galactoside-binding soluble 3 protein; lectin L-29; lectin, galactose binding, soluble 3; lectin, galactoside binding soluble 3; lectin, galactoside-binding, soluble, 3; lectin, galactoside-binding, soluble, 3 (galectin 3); LGALS2; Lgals3; MAC2; Mac-2; Mac-2 antigen
Gene Symbols LGALS3
Host Species Rabbit
Immunogen E.coli-derived Recombinant Human Galectin-3.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3958
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.