Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galectin-3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501729882 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9882 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9882 Supplier Proteintech Group Inc Supplier No. 149791AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The monoclonal antibody B2C10 reacts with galectin-3, a 30 kDa protein. Galectin-3 is a member of the galectin family. The protein is compose of three domains: a small amino-terminal domain, a carboxyl-terminal carbohydrate recognition domain (CRD) and amino-terminal domain containing repeating elements. Galectin-3 is normally distributed in epithelia of many organs and various inflammatory cells, including macrophages, as well as dendritic cells and Kupffer cells. The expression of this lectin is up-regulated during inflammation, cell proliferation, cell differentiation and through trans-activation by viral proteins. The expression is also affected by neoplastic transformation: up-regulated in certain types of lymphomas and thyroid carcinoma, while down-regulated in other types of maligcies, such as colon, breast, ovarian and uterine carcinomas.

Specifications

Antigen Galectin-3
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LGALS3
Gene Accession No. P08699, P16110, P17931
Gene Alias 35 kDa lectin, CBP 35, CBP35, Gal 3, GAL3, Galactose specific lectin 3, Galactoside binding protein, GALBP, Galectin 3, Galectin-3, GALIG, IgE binding protein, L 31, Laminin binding protein, Lectin L 29, LGALS2, LGALS3, Mac 2 antigen, MAC2
Gene Symbols Lgals3
Host Species Rabbit
Immunogen Galectin-3 Fusion Protein Ag6891
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16854, 3958, 83781
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.