Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galectin-3BP/MAC-2BP/LGALS3BP Antibody (OTI6B7), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271125AF700
Description
Galectin-3BP/MAC-2BP/LGALS3BP Monoclonal specifically detects Galectin-3BP/MAC-2BP/LGALS3BP in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
Galectin-3BP/MAC-2BP/LGALS3BP | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
LGALS3BP | |
Recombinant protein fragment corresponding to amino acids 19-300 of human Galectin-3BP/MAC-2BP/LGALS3BP (NP_005558) produced in E.coli. | |
0.1 mL | |
3959.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI6B7 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
90K, Basement membrane autoantigen p105, BTBD17B, galectin-3-binding protein, L3 antigen, Lectin galactoside-binding soluble 3-binding protein, lectin, galactoside-binding, soluble, 3 binding protein, M2BP, Mac-2 BP, Mac-2-binding protein, MAC2BP, MAC-2-BP, serum protein 90K, Tumor-associated antigen 90K | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction