Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galectin-3BP/MAC-2BP/LGALS3BP Rabbit anti-Human, Mouse, Rat, Clone: S03-5B8, Novus Biologicals™

Rabbit Monoclonal Antibody
$274.50 - $584.50
Specifications
Antigen | Galectin-3BP/MAC-2BP/LGALS3BP |
---|---|
Clone | S03-5B8 |
Dilution | Western Blot, ELISA 1:100-1:2000, Immunofluorescence 20 μg/mL |
Applications | Western Blot, ELISA, Immunofluorescence |
Classification | Monoclonal |
Description
Galectin-3BP/MAC-2BP/LGALS3BP Monoclonal antibody specifically detects Galectin-3BP/MAC-2BP/LGALS3BP in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
Galectin-3BP/MAC-2BP/LGALS3BP | |
Western Blot, ELISA 1:100-1:2000, Immunofluorescence 20 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
90K, Basement membrane autoantigen p105, BTBD17B, galectin-3-binding protein, L3 antigen, Lectin galactoside-binding soluble 3-binding protein, lectin, galactoside-binding, soluble, 3 binding protein, M2BP, Mac-2 BP, Mac-2-binding protein, MAC2BP, MAC-2-BP, serum protein 90K, Tumor-associated antigen 90K | |
Recombinant protein of human Galectin-3BP/MAC-2BP/LGALS3BP (Uniprot#: Q08380) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
S03-5B8 | |
Western Blot, ELISA, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer, Cell Biology, Cell Cycle and Replication | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
3959 | |
IgG | |
Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title