Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galectin-4 Antibody (MM0300-1D12), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212323AF405
Description
Galectin-4 Monoclonal specifically detects Galectin-4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Galectin-4 | |
Monoclonal | |
Alexa Fluor 405 | |
P56470 | |
LGALS4 | |
Human recombinant Galectin-4 | |
0.1 mL | |
Cancer | |
3960 | |
Store at 4C in the dark. | |
IgG2 |
Western Blot, Immunohistochemistry (Paraffin) | |
MM0300-1D12 | |
Western Blot, Immunohistochemistry-Paraffin | |
Antigen NY-CO-27, gal-4, GAL4, galectin 4, galectin-4, L-36 lactose-binding protein, L36LBP, Lactose-binding lectin 4, lectin, galactoside-binding, soluble, 4 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction